Organic reference standard, 2-Methylindole, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
41069-100MG
41069-100MGEA 146 EUR InStock
41069-100MG
Organic reference standard, 2-Methylindole, analytical standard, Supelco®
Standards Organische Standards
Regioselective synthesis of oxopyrrolidine analogs via iodine-catalysed Markovnikov addition reaction: Friedel-crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors, pyridyl-ethenyl-indoles as potential anticancer immunomodulators, preparation of plant-growth inhibitors, michael addition reactions, synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge