Mahlbecher „Comfort“ für PM 100, PM 100CM, PM 200, PM 400
Lieferant: Retsch GmbH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01.462.0239 01.462.0240
412-0083EA 567 EUR InStock
412-0083 412-0084
Mahlbecher „Comfort“ für PM 100, PM 100CM, PM 200, PM 400
Dosen Mahlbecher
Effective grinding can be achieved by the selection of grinding jars and grinding balls from a wide range of sizes and materials of construction. The ‘comfort’ grinding jars have several safety and design features that make them easy to use.
  • O-ring for gas-tight and dust-proof seal
  • Stainless steel protective jacket for jars that are not made of steel
  • Gap between flanges of jar and edge of lid for easy opening
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge