Rotary cone sample divider, LABORETTE 27
Lieferant: FRITSCH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27.1420.00
412-4191EA 3704 EUR InStock
412-4191
Rotary cone sample divider, LABORETTE 27
Probenteiler
The LABORETTE 27 sample divider divides samples with up to 3,000 dividing steps per minute centred over a rotating dividing cone up to 30 separate channels. Each individual sample with identical chemical and physical properties and with 99,9% accuracy.
  • Greatest division precision during sample division
  • Variable division ratios
  • Division of dry samples and suspensions without modification of the instrument
Order Now
PRODUKT
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge