Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Polysorbat 80 NF, Multikompendial, erfüllt die analytischen Spezifikationen von ChP, BP, JP, Ph. Eur.

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


Veränderungsmanagement-Kategorie= R

4125-04EA 404 EUR InStock
4125-04 4125-06 4125-03
Polysorbat 80 NF, Multikompendial, erfüllt die analytischen Spezifikationen von ChP, BP, JP, Ph. Eur.
Polysorbat 80
  • GMP manufactured product
  • Vegetable based

Suitable for use in the manufacture of parenteral dosage forms.


Melting Pt: –21 °C
Dichte: 1.062 g/cm³
Flash-Pt: 113 °C (geschlossener Tiegel)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00082107
CAS-Nummer: 9005-65-6
EINECS: 500-019-9
Order Now
Spezifikation Testergebnisse
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets N.F. Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Acid Value ≤ 2.0
NF - Hydroxyl Value 65 - 80
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Residue on Ignition ≤ 0.25 %
NF - Saponification Value 45 - 55
NF - Specific Gravity at 25°C 1.06 - 1.09
NF - Viscosity at 25°C, cSt 300 - 500
NF - Water (H₂O) ≤ 3.0 %
NF - Peroxide Value ≤ 10.0
NF - Ethylene Oxide ≤ 1 ppm
NF - Dioxane ≤ 10 ppm
NF - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
NF - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
NF - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
NF - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
NF - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
NF - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
NF - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
EP - Acid Value ≤ 2.0
EP - Total Ash ≤ 0.25 %
EP - Hydroxyl Value 65 - 80
EP - Identification A Passes Test
EP - Identification D Passes Test
EP - Peroxide Value ≤ 10.0
EP - Ethylene Oxide ≤ 1 ppm
EP - Dioxan ≤ 10 ppm
EP - Saponification Value 45 - 55
EP - Water (H₂O) ≤ 3.0 %
EP/BP - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
EP/BP - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
EP/BP - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
EP/BP - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
EP/BP - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
EP/BP - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
EP/BP - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
JP - Acid Value ≤ 2.0
JP - Composition of Fatty Acids - Myristic Acid ≤ 5.0 %
JP - Composition of Fatty Acids - Palmitic Acid ≤ 16.0 %
JP - Composition of Fatty Acids - Palmitoleic Acid ≤ 8.0 %
JP - Composition of Fatty Acids - Stearic Acid ≤ 6.0 %
JP - Composition of Fatty Acids - Oleic Acid ≥ 58.0 %
JP - Composition of Fatty Acids - Linoleic Acid ≤ 18.0 %
JP - Composition of Fatty Acids - Linolenic Acid ≤ 4.0 %
JP - Dioxane ≤ 10 ppm
JP - Ethylene Oxide ≤ 1 ppm
JP - Heavy Metals (as Pb) ≤ 20 ppm
JP - Hydroxyl Value, meq KOH/g 65 - 80
JP - Identification Passes Test
JP - Peroxide Value ≤ 10.0
JP - Residue on Ignition ≤ 0.25 %
JP - Water (H₂O) ≤ 3.0 %
JP - Saponification Value 45 - 55
ChP - Relative Density @ 20 °C 1.06 - 1.09
ChP - Viscosity @ 25 °C 350 - 550 mm2/s
ChP - Acid Value ≤ 2.0
ChP - Saponification Value 45 - 55
ChP - Hydroxyl Value 65 - 80
ChP - Iodine Value 18 - 24
ChP - Peroxide Value ≤ 10
ChP - Identification 1 Passes Test
ChP - Identification 2 Passes Test
ChP - Identification 3 Passes Test
ChP - Identification 4 Passes Test
ChP - Acidity or Alkalinity 5.0 - 7.5
ChP - Color Passes Test
ChP - Ethylene glycol ≤ 0.01 %
ChP - Diethylene glycol ≤ 0.01 %
ChP - Ethylene oxide ≤ 0.0001 %
ChP - Dioxane ≤ 0.001 %
ChP - Freezing Test Passes Test
ChP - Water ≤ 3.0 %
ChP - Residue on ignition ≤ 0.2 %
ChP - Heavy Metals ≤ 0.001 %
ChP - Arsenic (As) ≤ 0.0002 %
ChP - Fatty acid composition: oleic acid ≥ 58.0 %
ChP - Fatty Acid Composition: myristic acid ≤ 5.0 %
ChP - Fatty Acid Composition: palmitic acid ≤ 16.0 %
ChP - Fatty Acid Composition: palmitoleic acid ≤ 8.0 %
ChP - Fatty Acid Composition: stearic acid ≤ 6.0 %
ChP - Fatty Acid Composition: linoleic acid ≤ 18.0 %
ChP - Fatty Acid Composition: linolenic acid ≤ 4.0 %
Additional Tests - Endotoxin Concentration (EU/mL) ≤ 10
Microbiological - Total Aerobic Microbial Count (cfu/g) ≤ 100
Microbiological - Escherichia Coli
Microbiological - Pseudomonas aeruginosa
Microbiological - Salmonella
Microbiological - Staphylococcus aureus
Passes Test
Microbiological - Total Yeast and Mold Count (cfu/g) ≤ 50
Ethylene glycol ≤ 0.10 %
Diethylene glycol ≤ 0.10 %
Vegetable Based
This product utilizes ingredients of non-animal origin and non-peanut origin.
Suitable for use in the manufacture of parenteral dosage forms
Elemental Impurities (USP 232, EP 5.20)
Information on elemental impurities for this product is available on the associated Product
Only Class 2 (1,4 Dioxane, Ethylene Glycol) and Class 3 (acetic acid, 2-propanol) solvents are likely to be present
Class 2 solvents are below the Option 1 limits and any Class 3 solvent is <0.5%
Typical Oleic Acid Content, 77%

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge