Stabilization:
Stabilisiert mit BHT/Ionol/2,6-Di-tert-butyl-4-methylphenol
41400.02
41400.04
41400.09
41400.02EA
30.5
EUR
InStock
41400.02
41400.04
41400.09
Vinylene carbonate ≥97% stabilisiert
Vinylene carbonate
Stab. with BHT
Purity: 97+%
CAS No.: 872-36-6
Molecular Formula: C3H2O3
Formula Weight: 86.05
Storage Temperature: 2°C to 8°C
Flash Point: 72°(163°F)
Physical Form: Liquid
Density: 1.355
Refractive Index: 1.4210
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Melting Point: 19-22°
Boiling Point: 162-163°
MDL No.: MFCD00005380