41613.FF
41613.FFEA
721
EUR
InStock
41613.FF
Beryllium ≥99,5% (Metall-Basis), Folie, Dicke 0.05 mm (0.002 in)
Beryllium
Purity: 99.5%
CAS No.: 7440-41-7
Molecular Formula: Not vacuum tight
Physical Form: ≈0.058g/25x25mm, not certified pinhole free
Odour: Odourless