41738.06
41738.14
41738.22
41738.06EA
60.4
EUR
InStock
41738.06
41738.14
41738.22
Dioctylzinndichlorid ≥95%
Dioctylzinndichlorid
Purity: 95%
CAS No.: 3542-36-7
Molecular Formula: [CH3(CH2)7]2SnCl2
Formula Weight: 416.04
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Boiling Point: 175°/1mm
UN No.: 3261
MDL No.: MFCD00018091