41795.03
41795.06
41795.03EA
77
EUR
InStock
41795.03
41795.06
Wolfram(VI)ethanolat
Wolfram(VI)ethanolat
CAS No.: 62571-53-3
Molecular Formula: C12H30O6W
Formula Weight: 454.21
Physical Form: Powder/chunks
Sensitivity: Moisture Sensitive
Appearance: Off-white to brown
Melting Point: 150°
Solubility: Soluble in ethyl acetate
UN No.: 3181
MDL No.: MFCD00156526
Intermediate for sol-gel coatings