verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
41796.22
41796.36
41796.22EA
47.3
EUR
InStock
41796.22
41796.36
Bortrifluorid-Diethylether-Komplex (≥46,5% BF₃)
Bortrifluorid-Diethylether-Komplex
CAS No.: 109-63-7
Molecular Formula: C4H10BF3O
Formula Weight: 141.93
Flash Point: 47°(116°F)
Physical Form: Liquid
Density: 1.130
Refractive Index: 1.3440
Sensitivity: Moisture Sensitive
Odour: Acrid
Melting Point: -60°
Boiling Point: 125-126°
Beilstein Reference: 3909607
Merck Reference: 14,1350
UN No.: 2604
MDL No.: MFCD00013194
Packaged under Argon in resealable ChemSeal™ bottles