41958.14
41958.22
41958.14EA
178
EUR
InStock
41958.14
41958.22
Zinkselenid ≥99,99% (Metall-Basis)
Zinkselenid
Purity: 99.99%
CAS No.: 1315-09-9
Molecular Formula: ZnSe
Formula Weight: 144.33
Physical Form: -325 Mesh Powder
Density: 5.42
Melting Point: 1100°
Solubility: Insoluble in water. Decomposes in dilute HNO3
Merck Reference: 14,10156
UN No.: 3283
MDL No.: MFCD00011296