42040.06
42040.14
42040.06EA
38.8
EUR
InStock
42040.06
42040.14
Alizarinrot S (Natriumsalz)
Alizarinrot S (Natriumsalz)
CAS No.: 130-22-3
Molecular Formula: C14H7O7SNa
Formula Weight: 342.27
Physical Form: Powder
Appearance: Yellow to red
Beilstein Reference: 4117090
Merck Reference: 14,8573
MDL No.: MFCD00013049