42673.03
42673.06
42673.14
42673.03EA
69.1
EUR
InStock
42673.03
42673.06
42673.14
Germanium(IV)isopropoxid ≥99,9% (Metall-Basis)
Germanium(IV)isopropoxid
Purity: 99.9%
CAS No.: 21154-48-3
Molecular Formula: C12H28GeO4
Formula Weight: 308.94
Flash Point: 68°(154°F)
Physical Form: Liquid
Density: 1.025
Refractive Index: 1.414
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Alcohol-like
Boiling Point: 109°/30mm
MDL No.: MFCD00015339