Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Calcium phosphate tribasic, Pulver NF, FCC, Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


Veränderungsmanagement-Kategorie= R

4280-03EA 264.69 EUR InStock
4280-03 4280-24
Calcium phosphate tribasic, Pulver NF, FCC, Macron Fine Chemicals™
Calcium phosphate tribasic
Powder.


Formel: Ca₅HO₁₃P₃
MDL Number: MFCD00010904
CAS-Nummer: 1306-06-5
Order Now
Spezifikation Testergebnisse
Meets F.C.C. Requirements
GMP Manufactured Product
Meets N.F. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical 34.0 - 40.0 %
NF - Assay (Ca) 34.0 - 40.0 %
NF - Acid-Insoluble Substances ≤ 0.2 %
NF - Arsenic (As) ≤ 3 mg/kg
NF - Barium (Ba) Passes Test
NF - Carbonate (CO₃) Passes Test
NF - Chloride (Cl) ≤ 0.14 %
NF - Dibasic Salt and Calcium Oxide (ml) 13.0 - 14.3
NF - Fluoride (F) ≤ 0.0075 %
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Loss on Ignition ≤ 8.0 %
NF - Nitrate (NO₃) Passes Test
NF - Sulfate (SO₄) ≤ 0.8 %
NF - Water-Soluble Substances ≤ 0.5 %
FCC - Assay (Ca) 34.0 - 40.0 %
FCC - Arsenic (As) ≤ 3 mg/kg
FCC - Fluoride (F) ≤ 0.0075 %
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Loss on Ignition ≤ 8.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge