Unter Argon in Ampullen verpackt
42843.06
42843.14
42843.22
42843.06EA
61.2
EUR
InStock
42843.06
42843.14
42843.22
Magnesiumchlorid, ultra trocken ≥99,9% (Metall-Basis)
Magnesium chloride
Purity: 99.9%
CAS No.: 7786-30-3
Molecular Formula: Cl2Mg
Formula Weight: 95.22
Physical Form: Powder, Ampouled under argon
Density: 2.32
Appearance: White to yellow
Odour: Odourless
Melting Point: 714°
Merck Reference: 14,5662
MDL No.: MFCD00011106