42879.K2
42879.K2EA
109
EUR
InStock
42879.K2
Acetonitril ≥99,9%, Supergradienten-Qualität für HPLC, fernes UV
Acetonitril
Purity: 99.9+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odour: Pungent
Melting Point: -48° to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Suitable for gradient HPLC