43085.03
43085.06
43085.14
43085.03EA
199
EUR
InStock
43085.03
43085.06
43085.14
Palladium(II)chlorid (≥59,5% Pd) ≥99,999% (Metall-Basis) Pd ≥ 59.5%, Premion®
Palladium(II)chlorid
Purity: 99.999%
CAS No.: 7647-10-1
Molecular Formula: PdCl2
Formula Weight: 177.31
Physical Form: Powder
Density: 4.0
Appearance: Red-brown
Odour: Odourless
Melting Point: 675° dec.
Merck Reference: 14,6990
MDL No.: MFCD00003558