Homogeniser, high-throughput, HOHTDG
Lieferant: OHAUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30391396
431-0261EA 9380 EUR InStock
431-0261
Homogeniser, high-throughput, HOHTDG
Homogenisatoren Tischhomogenisatoren
The HT (High Throughput) lysing bead mill laboratory homogeniser provides efficient solutions for grinding, lysing, pulverising, mixing and homogenising in sample preparation applications.
  • Made with a polyurethane elastomer top housing, cycoloy base and a lexan lid
  • Flexibility to handle multiple samples and tube configurations
  • Five pre-set programs optimised for speed and time
  • Increased efficiency with high-throughput sample processing
  • Easy-to-read LED display allows the operator to view speed and time settings
  • Microprocessor-controlled unit includes sample clamp and hex key
An easy-to-load tray securely locks the microplate or tube in place. The high speed linear motion (31 mm) processes samples quickly. Ideal for DNA, RNA and protein analysis (from mammalian tissue, plant, soil, seed, fungi, bacteria, virus and other cells), cell lysing, PCR and gene expression.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge