Tissue grinders
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
196102 196105 196110 196115 196302 196305 196310 196315
432-1202EA 140 EUR InStock
432-1202 432-1205 432-1210 432-1215 432-1235 432-1240 432-1245 432-1250
Tissue grinders
Homogenisatoren Gewebehomogenisatoren
Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge