Cryolys® Accessory for Precellys 24 and Evolution
Lieferant: Bertin Instruments
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P000671-CLYS0A
432-3756EA 0 EUR InStock
432-3756
Cryolys® Accessory for Precellys 24 and Evolution
Boxen Kühl- und Gefrierboxen, Kühl- und Gefrierbeutel
Cryolys is a patented cooling option for Precellys homogenisers that prevents temperature increases during the homogenisation process, maintaining it within the physiological range, between 4 and 20 °C, depending on settings, strength and duration of homogenisation.
  • Cryolys prevents the temperature dependent degradation or denaturation of sensitive molecules like RNA or native proteins
  • Gives higher yields of intact target molecules during subsequent extraction and obtains better results during downstream analysis
Alternatively the Cryolys Evolution (432-0346) is available, which has been specifically designed for the Precellys Evolution homogeniser and only requires dry ice for operation.
Bestellinformation: The Cryolys models listed below require dry compressed air and liquid nitrogen or dry ice to operate.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge