Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Leucoberbelin Blue I 65% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

432199-100MG 432199-500MG
432199-100MGEA 98 EUR InStock
432199-100MG 432199-500MG
Leucoberbelin Blue I 65% (Farbstoffgehalt), Sigma-Aldrich®
Leucoberbelin Blue I
Leucoberbelin blue I has been used as a quencher in time-resolved fluorescence QRET technique for receptor-ligand interaction screening and to detect oxidized manganese species in Mn(II) (manganese) oxidation assays.

Leucoberbelin Blue I is a colourimetric reagent redox dye. It is a water-soluble leuco base also abbreviated as LBB. Upon oxidation, LBB forms a coloured complex which is stoichiometrically related to the oxidation state of the reagent.


Formel: C₂₃H₂₆N₂O₃S
MW: 410.53 g/mol
Melting Pt: >300 °C (Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00217522
CAS-Nummer: 52748-86-4
UN: 3261
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d85579b5f44f1e6bea105de8cbbd761aa41190d&vid=5d85579b5f44f1e6bea105de8cbbd761aa41190d&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance Light blue powder
Infrared spectrum Conforms to structure and standard as Illustrated on page 4132C of edition I, Volume 3 of 'the aldrich library of FT-IR Spectra'.
Elemental analysis Carbon 42.4% (minimum), Nitrogen 4.3% (minimum)
Thin-layer Consistent with control

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge