43225.18
43225.A1
43225.30
43225.18EA
33.5
EUR
InStock
43225.18
43225.A1
43225.30
Chinolin ≥98%
Chinolin
Purity: 98%
CAS No.: 91-22-5
Molecular Formula: C9H7N
Formula Weight: 129.16
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.090
Refractive Index: 1.6260
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colourless to pale yellow
pH: 7.3
Melting Point: -15°
Boiling Point: 237°
Beilstein Reference: 107477
Merck Reference: 14,8068
UN No.: 2656
MDL No.: MFCD00006736