43488.30
43488.14
43488.A1
43488.30EA
91.4
EUR
InStock
43488.30
43488.14
43488.A1
Arsen(III)oxid ≥99,5% (Metall-Basis)
Arsen(III)oxid
Purity: 99.5%
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: -100 Mesh Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Solubility: Slightly soluble in cold water. Soluble in boiling water, HCl, alkali hydroxide. Insoluble in alcohol, chloroform, and ether
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433