43491.06
43491.03
43491.06EA
289
EUR
InStock
43491.06
43491.03
1-(2-Bromethyl)naphthalin ≥97%
1-(2-Bromoethyl)naphthalene
Purity: 97%
CAS No.: 13686-49-2
Molecular Formula: C12H11Br
Formula Weight: 235.13
Flash Point: 110°(230°F)
Physical Form: Liquid
Boiling Point: 237-238°1.65
MDL No.: MFCD00037737