43701.22
43701.14
43701.22EA
127
EUR
InStock
43701.22
43701.14
Bisacrylamid ≥99%
Bisacrylamid
Purity: 99+%
CAS No.: 110-26-9
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Physical Form: Powder
Density: 1.235
Appearance: White
Melting Point: 300°
Beilstein Reference: 1706297
MDL No.: MFCD00008625