43702.03
43702.06
43702.03EA
1245
EUR
InStock
43702.03
43702.06
Rhodium(III)chlorid Hydrat ≥99,99% (Metall-Basis), Premion®
Rhodium(III)chlorid Hydrat
Purity: 99.99%
CAS No.: 20765-98-4
Molecular Formula: RhCl3.xH2O
Formula Weight: 209.26(anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Dark red
Melting Point: 100° dec.
Merck Reference: 14,8188
MDL No.: MFCD00149839