Multi-element standard solution 6 for ICP, TraceCERT®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
43843-100ML
43843-100MLEA 155 EUR InStock
43843-100ML
Multi-element standard solution 6 for ICP, TraceCERT®
Standards Kationischer Standards Mehrelement-Standards
This standard solution is intended for use as a certified reference material for ICP/AAS.
Multi element standards for Al, Sb, Ba, Pb, B, Ca, Cd, Cr, Co, Fe, K, Cu, Li, Mg, Mn, Mo, Na, Ni, P, Si, Ti, V and Zn at 100 mg/ml in 5% nitric acid and ≤0,1% hydrofluoric acid.
Zertifizierungen: Accredited to ISO 17034 and ISO/IEC 17025.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge