Organic reference standard, 2-(4-tert-Butylbenzyl)propionaldehyde, analytical standard, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
43884-10MG
43884-10MGEA 53.6 EUR InStock
43884-10MG
Organic reference standard, 2-(4-tert-Butylbenzyl)propionaldehyde, analytical standard, Supelco®
Standards Organische Standards
2-(4-tert-Butylbenzyl)propionaldehyde is categorised under the synthetic fragrance class of compounds widely utilised in consumer products such as perfumes, after shave lotions, cosmetics and more.
2-(4-tert-Butylbenzyl)propionaldehyde can be used as a reference standard for the determination of 2-(4-tert-butylbenzyl)propionaldehyde in: Human urine samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionisation (ESI) source and multiple reaction monitoring (MRM) mode of detection, deodorants and air fresheners by sonication extraction coupled with gas chromatography-mass spectrometry (GC-MS), scented consumer products by GC-MS as well as GC in combination with olfactometry.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e951b214aa12013eececb3b797af9a505ad2fc4c&vid=e951b214aa12013eececb3b797af9a505ad2fc4c&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge