Electronic overhead stirrers, MINISTAR 80 control
Lieferant: IKA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
25001990
441-0469EA 1729 EUR InStock
441-0469
Electronic overhead stirrers, MINISTAR 80 control
Rührer Rührwerke
This overhead stirrer is ideal for special applications. Combining high performance with excellence, the units require minimum space.
  • Enclosed in hardened glass
  • Fast, response display for maximum visibility and chemical resistance
  • Clear display shows all essential information at a glance
  • Viscosities up to 60,000 mPas and volumes of up to 50000 ml
  • Speed is continuously adjustable between 0/30 – 500 rpm
Zertifizierungen: IP protection class according to DIN EN 60529: IP 54
Bestellinformation: Supplied without accessories, a wide range of accessories are available to order separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge