Electronic overhead stirrers, Hei-TORQUE core
Lieferant: Heidolph Instruments GmbH & Co.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
501-60410-00
441-0471EA 1081 EUR InStock
441-0471
Electronic overhead stirrers, Hei-TORQUE core
Rührer Rührwerke
Hei-TORQUE Core's compact design allows for integration in closed systems, such as fume hoods, reactors, or production systems. Suitable for low- to medium-viscosity media up to 25 L. The large diameter of the chuck allows you to use even large impellers and tools. This facilitates a wide variety of applications, such as homogenisation, dispersing, dissolving of agglomerates, and more.
  • Control knob for rotation speed, pushing starts or stops the function
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge