unter Argon abgefüllt
44132.03
44132.06
44132.14
44132.03EA
99.9
EUR
InStock
44132.03
44132.06
44132.14
N,N'-Bis(acryloyl)cystamine ≥97%
N,N'-Bis(acryloyl)cystamine
Purity: 98%
CAS No.: 60984-57-8
Molecular Formula: C10H16N2O2S2
Formula Weight: 260.37
Storage Temperature: -30°C to -10°C
Physical Form: Powder, Packaged under argon
Appearance: White
Melting Point: 123-124°
MDL No.: MFCD00036225
Applications: Used as a reversible cross-linker for polyacrylamide gels