Grease, high vacuum, Dow Corning®
Lieferant: Alfa Aesar
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
44224.KT
44224.KTEA 82.2 EUR InStock
44224.KT
Grease, high vacuum, Dow Corning®
Schmiermittel
High-vacuum grease is used as an anti-seize grease in ground glass joints and to lubricate synthetic and silicone rubber gaskets. Also used to lubricate water-treating equipment, laboratory equipment, lubricate O-rings on binoculars and telescopes and to prevent fogging of delicate lenses.
  • Recommended for vacuums reaching between 1×10-5 and 1×10-6 mmHg
  • Immiscible with water, methanol, ethanol, acetone, acetone, glycol and glycerine
  • Incompatible with oxidising agents
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge