44345.04
44345.09
44345.18
44345.04EA
72.8
EUR
InStock
44345.04
44345.09
44345.18
Cobalt(II)acetat Tetrahydrat ≥99,999% (Metall-Basis)
Cobalt(II)acetat Tetrahydrat
Purity: 99.999%
CAS No.: 6147-53-1
Molecular Formula: C4H6CoO4.4H2O
Formula Weight: 249.08 (177.02anhy)
Physical Form: Crystalline
Density: 1.71
Refractive Index: 1.542
Sensitivity: Hygroscopic
Appearance: Red
Odour: Odourless
Melting Point: ca 140° -4H2O
Beilstein Reference: 3731397
Merck Reference: 14,2433
UN No.: 3077
MDL No.: MFCD00149569