Attachments for vortex mixers, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H370800020
444-0091EA 376 EUR InStock
444-0091
Attachments for vortex mixers, SP Bel-Art
Gestelle Reagenzglasgestelle
Vortexer attachment for microcentrifuge tubes can hold 1,5 ml tubes in an 8 or 20 capacity disk to carry out simultaneous vortexing. The contents present in the tube are mixed by holding the shaft against the rubber pad of a vortexer.
  • Made of PP
  • Mix up to 20 tubes at single time
  • Shaft length: 165 mm
  • Ball handle Ø: 254 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge