Agitateur à mouvement circulaire
Lieferant: GERHARDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11-0002
444-0166EA 0 EUR InStock
444-0166
Agitateur à mouvement circulaire
Schüttler Kreisschüttler
Separate LED displays allows shaking time and speed to be called up and changed during any procedure.
  • Nine different shaking programmes, each with nine steps per programme for speed and time
  • Rubber coated shaking platform
Timer: 0 to 99 h 59 min 59 s or continuous mode.
Bestellinformation: Supplied with 4×150 mm vertical rods for different attachments.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge