Kreisförmig vibrierender Plattformschüttler, PSU-10i
Lieferant: GRANT INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PSU-10I
444-0933EA 978 EUR InStock
444-0933
Kreisförmig vibrierender Plattformschüttler, PSU-10i
Schüttler Kreisschüttler
Orbital flatbed shaker, with LCD display of both set and actual speed, plus set and elapsed time. Available with a choice of four interchangeable accessory platforms, to enable use with a wide range of vessels, including bottles, flasks and beakers, dishes, boxes and petri dishes. An audible signal accompanies automatic switch-off to indicate the completion of a set time period. The soft start feature ensures a smooth ramp up of the shaking speed.
  • Variable speed control
  • Continuous or timer operation from 1 min to 96 h with automatic switch-off
  • Can be used at +4 to +40 °C, in an incubator or cold room
  • Low voltage power supply enables safe cold room operation
* Maximum speed is dependent on load.
Bestellinformation: Supplied without platform, required accessories must be ordered separately.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge