Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Orbital shaker platform, Bel-Art ProCulture, SP Bel-Art

Lieferant: Bel-Art Products, a Part of SP

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

F370410000
444-2160EA 942 EUR InStock
444-2160
Orbital shaker platform, Bel-Art ProCulture, SP Bel-Art
Schüttler Kreisschüttler
The orbital shaker platform turns a standard magnetic stirrer into an orbital shaking platform saving the expense and space requirements of additional lab equipment.

  • Economical
  • Compact design
  • Shake and stir; rotating orbit diameter of 21.7 mm (7/8")
  • Shaker adjustment is controlled by setting the magnetic stirrer to the desired speed
  • Base incorporates built-in hand holds for lifting and carrying
  • Supports up to 6.8 kg (15 lb)

Platform protects the operating mechanism from spills and has a non-skid surface to prevent containers from sliding.

Zertifizierungen: U.S. Patent 5,409,312.

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge