J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
Lieferant: Avantor
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4463-02EA 389.58 EUR InStock
4463-02 4463-04
J.T.Baker® BAKER-flex®, Precoated flexible TLC sheet
TLC-Platten
Ready-to-use in thin layer chromatography applications, Baker-flex® precoated flexible plastic sheet have a hard, abrasion-resistant coating and solvent-resistant substrate for rapid, clean separations.
The sheets can be written on and cut to size. They are available with or without fluorescent indicator.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge