44816.14
44816.22
44816.14EA
52.9
EUR
InStock
44816.14
44816.22
Quecksilber(II)bromid ACS
Quecksilber(II)bromid
CAS No.: 7789-47-1
Molecular Formula: Br2Hg
Formula Weight: 360.40
Physical Form: Powder
Density: 6.05
Sensitivity: Light Sensitive
Appearance: White
Odour: Odourless
Melting Point: 236°
Boiling Point: 322-325° subl.
Solubility: Soluble in hot alcohol, methanol, HCl, HBr
Merck Reference: 14,5875
UN No.: 1634
MDL No.: MFCD00011040