verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
44913.5Y
44913.AE
44913.K2
44913.5YEA
130
EUR
InStock
44913.5Y
44913.AE
44913.K2
N,N-Dimethylacetamid, wasserfrei ≥99,8%
N,N-Dimethylacetamid
Purity: 99.8%
CAS No.: 127-19-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 0.940
Refractive Index: 1.4385
Appearance: Colourless
Odour: Ammonia-like
Melting Point: -20°
Boiling Point: 165-166°
Beilstein Reference: 1737614
Merck Reference: 14,3227
MDL No.: MFCD00008686