45007.14
45007.22
45007.36
45007.14EA
39.3
EUR
InStock
45007.14
45007.22
45007.36
Eisen(III)oxid, NanoArc®, industriell
Eisen(III)oxid
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: 20-40nm APS Powder, S.A. 30-60m2/g
Density: 5.24
Appearance: Red-brown
Odour: Odourless
Melting Point: 1565°
MDL No.: MFCD00011008
NanoArc® industrial iron oxide is a general purpose gamma-phase material suitable for use in catalytic applications such as ammonia synthesis and dehydrogenation reactions. Manufactured by growing nanocrystals rapidly in..