45047.A3
45047.36
45047.A3EA
686
EUR
InStock
45047.A3
45047.36
Antimon ≥99,8% (Metall-Basis), gebrochener Stab, Länge various, Ø 12.7 mm (0.50 in)
Antimony
Purity: 99.8%
CAS No.: 7440-36-0
Physical Form: Broken pieces from cast rod
Odour: Odourless
MDL No.: MFCD00134030