45108.KF
45108.KFEA
212
EUR
InStock
45108.KF
Tellur ≥99,9% (Metall-Basis), Barren/Knopf, Ø ∼38 mm (1.5 in), Dicke 6 mm (0.24 in)
Tellurium
Purity: 99.9%
CAS No.: 13494-80-9
Physical Form: ≈30g/piece
Appearance: Silver
Odour: Odourless
MDL No.: MFCD00134062