Reihenheizgerät, EV16
Lieferant: GERHARDT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10-0006
453-1996EA 0 EUR InStock
453-1996
Reihenheizgerät, EV16
Heizgeräte Heizplatten
With six plates in a row. For temperatures up to 425 °C.
  • With variable output control
  • Stainless steel cover with mounting hole for support rod, M10 thread
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge