Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ceran® hotplates, LHG

Lieferant: GESTIGKEIT HARRY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

11SR-230V 33SR-400V 44SR-230V 11A 22A 33A 44A 4A 11SR 22SR 33SR-220V 44SR 4SR
453-0033EA 0 EUR InStock
453-0033 453-0035 GEST44SR-230V 453-2111 453-2112 453-2113 453-2114 453-2115 453-2121 453-2122 453-2123 453-2124 GEST4SR
Ceran® hotplates, LHG
Heizgeräte Heizplatten
Glass ceramic heating plate, which is extremely resistant to breakage and changes of temperature, will not distort, is permeable to ultra-violet light and also highly acid-resistant.

  • Uniform heating across entire surface
  • Insulated stainless steel housing
  • Temperature control by an NiCr-Ni sensor (regulation from 50 to 500 °C)

Four feet, individually adjustable, ensure wobble-free, horizontal stability.

Achtung! Bitte beachten: Note: models 33 A/SR and 44 A/SR requires a 3-phase (400 V) supply.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge