45462.09
45462.04
45462.09EA
292
EUR
InStock
45462.09
45462.04
Blei(II)sulfid ≥99,995% (Metall-Basis)
Blei(II)sulfid
Purity: 99.995%
CAS No.: 1314-87-0
Molecular Formula: PbS
Formula Weight: 239.25
Physical Form: Lump
Density: 7.5
Refractive Index: 3.921
Odour: Odourless
Melting Point: 1114°
Merck Reference: 14,5421
UN No.: 3077
MDL No.: MFCD00016280