45504.06
45504.14
45504.06EA
213
EUR
InStock
45504.06
45504.14
Kupfer ≥99,9% (Metall-Basis), Nano-Pulver APS 20-50 nm
Copper
Purity: 99.9%
CAS No.: 7440-50-8
Molecular Formula: Cu(core)/CuO(shell)
Physical Form: CuO layer approx. 2nm
Appearance: Black
Odour: Odourless
UN No.: 3089
MDL No.: MFCD00010965