45548.88
45548.ME
45548.88EA
594
EUR
InStock
45548.88
45548.ME
Kaliumtetrabromoplatinat(II) ≥99,9% (Metall-Basis)
Potassium tetrabromoplatinate(II)
Purity: 99.9%
CAS No.: 13826-94-3
Molecular Formula: Br4K2Pt
Formula Weight: 592.90
Physical Form: Crystalline
Melting Point: 300°
MDL No.: MFCD00011376