45565.04
45565.ME
45565.04EA
783
EUR
InStock
45565.04
45565.ME
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
(Pentamethylcyclopentadienyl)iridium(III) dichloride dimer
CAS No.: 12354-84-6
Molecular Formula: C20H30Cl4Ir2
Formula Weight: 796.67
MDL No.: MFCD00075435