45612.14
45612.22
45612.36
45612.14EA
90.9
EUR
InStock
45612.14
45612.22
45612.36
Hafnium(IV)chlorid ≥99,9% (Metall-Basis) (max. 0,5% Zr)
Hafnium(IV)chlorid
Purity: 99.9%
CAS No.: 13499-05-3
Molecular Formula: Cl4Hf
Formula Weight: 320.30
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White
Odour: Irritating
Melting Point: 320° subl.
Merck Reference: 14,4588
UN No.: 3260
MDL No.: MFCD00011034