Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Lithiumperchlorat Trihydrat ACS

Lieferant: ALFA AESAR

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

45923.22 45923.36
45923.22EA 77.6 EUR InStock
45923.22 45923.36
Lithiumperchlorat Trihydrat ACS
Lithiumperchlorat Trihydrat
Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in diels-alder reactions.

Lithium perchlorate trihydrate is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the baylis-hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.


Formel: ClLiO₄·3H₂O
Dichte: 1,84 g/cm³
Storage Temperature: Raumtemperatur
MDL Number: MFCD00149765
CAS-Nummer: 13453-78-6
EINECS: 232-237-2
UN: 1481
ADR: 5.1,II
Merck Index: 14,05539
Order Now
Spezifikation Testergebnisse
Assay (Unspecified) 63.0 - 68.0% LiClO₄
Heavy Metals (ICP-OES) 5 ppm
Impurity Content, Cl 0.003%
Impurity Content, Fe 5 ppm
Impurity Content, SO₄ 0.001%
Insoluble Material 0.005%
pH 6.0 - 7.5

 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge